Kalium(2-formylphenyl)trifluoroborat 96%
Lieferant: Apollo Scientific
PC1722-1G
PC1722-5G
APOSPC1722-1GEA
67.2
EUR
APOSPC1722-1G
APOSPC1722-5G
Kalium(2-formylphenyl)trifluoroborat 96%
Kalium(2-formylphenyl)trifluoroborat
Formel:
C₇H₅BF₃KO Schmelzpunkt: 220…224 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD02093979 CAS-Nummer: 192863-39-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...