Kalium(3,5-difluorphenyl)trifluoroborat 96%

Lieferant: Apollo Scientific

Synonyme: (3,5-Difluorphenyl)trifluorborsäure Kaliumsalz

PC1724-5G
APOSPC1724-5GEA 56.7 EUR
APOSPC1724-5G
Kalium(3,5-difluorphenyl)trifluoroborat 96%
Kalium(3,5-difluorphenyl)trifluoroborat
Formel: C₆H₃BF₅K
Molecular Weight: 219,99 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06659939
CAS-Nummer: 267006-26-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR