Kalium(3,4-dichlorphenyl)trifluoroborat 98%
Lieferant: Apollo Scientific
|
Achtung
|
PC1726-1G
PC1726-5G
PC1726-25G
APOSPC1726-1GEA
0
EUR
APOSPC1726-1G
APOSPC1726-5G
APOSPC1726-25G
Kalium(3,4-dichlorphenyl)trifluoroborat 98%
Kalium(3,4-dichlorphenyl)trifluoroborat
Formel:
C₆H₃BCl₂F₃K Schmelzpunkt: 178…180 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD04115763 CAS-Nummer: 850623-68-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...