Kalium(4-bromphenyl)trifluoroborat 96%

Lieferant: Apollo Scientific
Achtung

Synonyme: (4-Bromphenyl)trifluorborsäure Kaliumsalz

PC1749-25G
APOSPC1749-25GEA 75.4 EUR
APOSPC1749-25G
Kalium(4-bromphenyl)trifluoroborat 96%
Kalium(4-bromphenyl)trifluoroborat
Formel: C₆H₄BBrF₃K
Molecular Weight: 262.91 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02093978
CAS-Nummer: 374564-35-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR