Kalium(3,5-bis(trifluormethyl)phenyl)trifluoroborat 96%

Lieferant: Apollo Scientific

PC1769-25G PC1769-5G
APOSPC1769-25GEA 642 EUR
APOSPC1769-25G APOSPC1769-5G
Kalium(3,5-bis(trifluormethyl)phenyl)trifluoroborat 96%
Kalium(3,5-bis(trifluormethyl)phenyl)trifluoroborat
Formel: C₈H₃BF₉K
Lagertemperatur: Raumtemperatur
MDL: MFCD03701625
CAS-Nummer: 166328-09-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR