3-Carboxy-5-fluorphenylboronsäure 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-Carboxy-5-fluorobenzeneboronic acid, 3-Borono-5-fluorobenzoic acid
PC1770-5G
APOSPC1770-5GEA
127
EUR
APOSPC1770-5G
3-Carboxy-5-fluorphenylboronsäure 98%
3-Carboxy-5-fluorphenylboronsäure
Formel:
C₇H₆BFO₄ Schmelzpunkt: 112…116 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD07363791 CAS-Nummer: 871329-84-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...