4-Ethoxy-3-(trifluormethyl)phenylboronsäure 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Ethoxy-3-(trifluoromethyl)benzeneboronic acid, 4-Ethoxy-3-trifluoromethylphenylboronic acid

PC1773-1G
APOSPC1773-1GEA 63.8 EUR
APOSPC1773-1G
4-Ethoxy-3-(trifluormethyl)phenylboronsäure 98%
4-Ethoxy-3-(trifluormethyl)phenylboronsäure
Formel: C₉H₁₀BF₃O₃
Molecular Weight: 233.98 g/mol
Schmelzpunkt: 186…190 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD07363789
CAS-Nummer: 871329-83-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR