3-Fluor-(5-methoxycarbonyl)phenylboronsäure 96%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Fluoro-5-methoxycarbonylphenylboronic Acid, 3-Fluoro-5-(methoxycarbonyl)benzeneboronic acid

PC1774-5G
APOSPC1774-5GEA 149 EUR
APOSPC1774-5G
3-Fluor-(5-methoxycarbonyl)phenylboronsäure 96%
3-Fluor-(5-methoxycarbonyl)phenylboronsäure
Formel: C₈H₈BFO₄
Molecular Weight: 197.96 g/mol
Schmelzpunkt: 138…142 °C
Lagertemperatur: Kühlschrank
MDL: MFCD07363749
CAS-Nummer: 871329-62-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR