4-Fluor-3-(trifluormethyl)phenylboronsäure 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Fluoro-3-(trifluoromethyl)benzeneboronic acid

PC1779-5G PC1779-1G PC1779-25G
APOSPC1779-5GEA 24.8 EUR
APOSPC1779-5G APOSPC1779-1G APOSPC1779-25G
4-Fluor-3-(trifluormethyl)phenylboronsäure 98%
4-Fluor-3-(trifluormethyl)phenylboronsäure
Formel: C₇H₅BF₄O₂
Molecular Weight: 207.92 g/mol
Schmelzpunkt: 170…174 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD07363787
CAS-Nummer: 182344-23-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR