4-Fluoro-1-indanone 95+%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Fluoro-2,3-dihydro-1H-inden-1-one
PC1783-5G
APOSPC1783-5GEA
62.5
EUR
APOSPC1783-5G
4-Fluoro-1-indanone 95+%
4-Fluoro-1-indanone
Formel:
C₉H₇FO Molecular Weight: 150.152 g/mol Schmelzpunkt: 79…80 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00797829 CAS-Nummer: 699-99-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...