4-Chlor-3-fluorphenylacetonitril 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-(4-Chloro-3-fluorophenyl)acetonitrile
PC1803-1G
APOSPC1803-1GEA
21.9
EUR
APOSPC1803-1G
4-Chlor-3-fluorphenylacetonitril 98%
4-Chlor-3-fluorphenylacetonitril
Formel:
C₈H₅ClFN Lagertemperatur: Raumtemperatur |
MDL:
MFCD04115860 CAS-Nummer: 251570-03-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...