5,7-Bis(trifluoromethyl)-4-chloroquinoline 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Chloro-5,7-bis(trifluoromethyl)quinoline
PC1812-1G
APOSPC1812-1GEA
142
EUR
APOSPC1812-1G
5,7-Bis(trifluoromethyl)-4-chloroquinoline 98%
5,7-Bis(trifluoromethyl)-4-chloroquinoline
Formel:
C₁₁H₄ClF₆N Lagertemperatur: Raumtemperatur |
CAS-Nummer:
234450-34-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...