1-[(2-Chloroethyl)sulphonyl]-3-(trifluoromethyl)benzene 97%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 2-Chloroethyl 3-[(trifluoromethyl)phenyl] sulphone

PC1813-10MG
APOSPC1813-10MGEA 65.4 EUR
APOSPC1813-10MG
1-[(2-Chloroethyl)sulphonyl]-3-(trifluoromethyl)benzene 97%
1-[(2-Chloroethyl)sulphonyl]-3-(trifluoromethyl)benzene
Formel: C₉H₈ClF₃O₂S
Molecular Weight: 272,68 g/mol
Schmelzpunkt: 52…55 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00068016
CAS-Nummer: 175203-09-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR