3-Chlor-4-fluoranilin 98%
Lieferant: Apollo Scientific
|
Gefahr
|
PC1820-100G
PC1820-500G
PC1820-1KG
APOSPC1820-100GEA
0
EUR
APOSPC1820-100G
APOSPC1820-500G
APOSPC1820-1KG
3-Chlor-4-fluoranilin 98%
3-Chlor-4-fluoranilin
Intermediate for the synthesis of quinoline antibiotic norfloxacin. For an efficient synthesis see: J. Chem. Soc., Chem. Comm., 1339 (1995)
Formel:
C₆H₅ClFN Siedepunkt: 227…228 °C Schmelzpunkt: 42…47 °C Dichte: 1,42 g/cm³ (25 °C) Flammpunkt: 149 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007767 CAS-Nummer: 367-21-5 EINECS: 206-682-8 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...