4-Chlor-2-fluorbenzaldehyd 97%

Lieferant: Apollo Scientific
Achtung

PC1831-25G PC1831-100G PC1831-500G
APOSPC1831-25GEA 26.1 EUR
APOSPC1831-25G APOSPC1831-100G APOSPC1831-500G
4-Chlor-2-fluorbenzaldehyd 97%
4-Chloro-2-fluorobenzaldehyde
Formel: C₇H₄ClFO
Molecular Weight: 158.56 g/mol
Schmelzpunkt: 58…61 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00143282
CAS-Nummer: 61072-56-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR