4-Ethyl-5-fluoropyrimidine 95%
Lieferant: Apollo Scientific
PC1837-5G
APOSPC1837-5GEA
112
EUR
APOSPC1837-5G
4-Ethyl-5-fluoropyrimidine 95%
4-Ethyl-5-fluoropyrimidine
Formel:
C₆H₇FN₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD06657566 CAS-Nummer: 137234-88-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...