Fluorocholine chloride

Lieferant: Apollo Scientific
Achtung

Synonyme: N-(Fluoromethyl)-2-hydroxy-N,N-dimethylethanaminium chloride

PC1838-250MG
APOSPC1838-250MGEA 30.6 EUR
APOSPC1838-250MG
Fluorocholine chloride
Fluorocholine chloride
Formel: C₅H₁₃ClFNO
Molecular Weight: 157.62 g/mol
Schmelzpunkt: 184…185 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD07995053
CAS-Nummer: 459424-38-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR