6-Chloro-5-fluoro-2-mercaptobenzimidazole 97%

Lieferant: Apollo Scientific

Synonyme: 6-Chloro-5-fluorobenzimidazole-2-thiol

PC1860G-1G
APOSPC1860G-1GEA 239 EUR
APOSPC1860G-1G
6-Chloro-5-fluoro-2-mercaptobenzimidazole 97%
6-Chloro-5-fluoro-2-mercaptobenzimidazole
Formel: C₇H₄ClFN₂S
Molecular Weight: 202,64 g/mol
Schmelzpunkt: 298…301 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00052490
CAS-Nummer: 142313-30-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR