3-Chlor-4-fluorbenzoesäure 98%
Lieferant: Apollo Scientific
|
Achtung
|
PC1861D-100G
PC1861D-10G
PC1861D-25G
PC1861D-500G
APOSPC1861D-100GEA
69.6
EUR
APOSPC1861D-100G
APOSPC1861D-10G
APOSPC1861D-25G
APOSPC1861D-500G
3-Chlor-4-fluorbenzoesäure 98%
3-Chloro-4-fluorobenzoic acid
Formel:
C₇H₄ClFO₂ Molecular Weight: 174.56 g/mol Siedepunkt: 273 Schmelzpunkt: 133…136 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00042477 CAS-Nummer: 403-16-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...