2-Chlor-6-fluorbenzonitril 98%

Lieferant: Apollo Scientific
Achtung

PC1863-25G
APOSPC1863-25GEA 21.9 EUR
APOSPC1863-25G
2-Chlor-6-fluorbenzonitril 98%
2-Chlor-6-fluorbenzonitril

Treatment with nucleophiles e.g alkoxides can selectively displace the fluorine atom: Synth. Commun., 621 (1984)

Formel: C₇H₃ClFN
Molecular Weight: 155,56 g/mol
Siedepunkt: 104…105 °C/11mmHg
Schmelzpunkt: 55…60 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00001780
CAS-Nummer: 668-45-1
EINECS: 211-571-2
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR