4-Chloro-6-fluoro-2H-benzopyran-3-carboxaldehyde 97%

Lieferant: Apollo Scientific
Achtung

PC1863H-250MG
APOSPC1863H-250MGEA 21.9 EUR
APOSPC1863H-250MG
4-Chloro-6-fluoro-2H-benzopyran-3-carboxaldehyde 97%
4-Chloro-6-fluoro-2H-benzopyran-3-carboxaldehyde
Formel: C₁₀H₆ClFO₂
Molecular Weight: 212,61 g/mol
Schmelzpunkt: 84…86 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00084953
CAS-Nummer: 105799-69-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR