2-Chlor-5-fluorphenol 98%
Lieferant: Apollo Scientific
|
Gefahr
|
PC1880A-5G
APOSPC1880A-5GEA
21.9
EUR
APOSPC1880A-5G
2-Chlor-5-fluorphenol 98%
2-Chlor-5-fluorphenol
Formel:
C₆H₄ClFO Molecular Weight: 146,55 g/mol Siedepunkt: 183…185 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00042524 CAS-Nummer: 3827-49-4 UN: 3265 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...