2-Fluorobenzylmagnesium chloride 0,25 M in Diethylether

Lieferant: Apollo Scientific
Gefahr

PC1897-100ML
APOSPC1897-100MLEA 0 EUR
APOSPC1897-100ML
2-Fluorobenzylmagnesium chloride 0,25 M in Diethylether
2-Fluorobenzylmagnesium chloride
Formel: C₇H₆ClFMg
Lagertemperatur: Raumtemperatur
MDL: MFCD01319900
CAS-Nummer: 120608-58-4
UN: 1993
ADR: 3,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR