1-[4-(Trifluoromethyl)phenyl]but-1-en-3-one 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC1921-1G
PC1921-5G
PC1921-250MG
APOSPC1921-1GEA
0
EUR
APOSPC1921-1G
APOSPC1921-5G
APOSPC1921-250MG
1-[4-(Trifluoromethyl)phenyl]but-1-en-3-one 97%
1-[4-(Trifluoromethyl)phenyl]but-1-en-3-one
Formel:
C₁₁H₉F₃O Siedepunkt: 118 °C/4mmHg Lagertemperatur: Raumtemperatur |
MDL:
MFCD00052848 CAS-Nummer: 80992-93-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...