6-Fluoro-2-methylquinoline-3-carboxylic acid

Lieferant: Apollo Scientific

PC1927-1G
APOSPC1927-1GEA 441 EUR
APOSPC1927-1G
6-Fluoro-2-methylquinoline-3-carboxylic acid
6-Fluoro-2-methylquinoline-3-carboxylic acid
Formel: C₁₁H₈FNO₂
Molecular Weight: 205.19 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03085774
CAS-Nummer: 461026-47-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR