6-Chloro-7-hydroxy-4-(trifluoromethyl)coumarin 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 6-Chloro-7-hydroxy-4-(trifluoromethyl)-2H-chromen-2-one

PC1931C-500MG
APOSPC1931C-500MGEA 21.9 EUR
APOSPC1931C-500MG
6-Chloro-7-hydroxy-4-(trifluoromethyl)coumarin 97%
6-Chloro-7-hydroxy-4-(trifluoromethyl)coumarin
Formel: C₁₀H₄ClF₃O₃
Molecular Weight: 264,59 g/mol
Schmelzpunkt: 178…182 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00068095
CAS-Nummer: 119179-66-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR