4'-Chloro-3'-methyl-2-nitro-4-(trifluoromethyl)diphenyl ether 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC1936P-1G
APOSPC1936P-1GEA
161
EUR
APOSPC1936P-1G
4'-Chloro-3'-methyl-2-nitro-4-(trifluoromethyl)diphenyl ether 97%
4'-Chloro-3'-methyl-2-nitro-4-(trifluoromethyl)diphenyl ether
Formel:
C₁₄H₉ClF₃NO₃ Schmelzpunkt: 57…59 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00052259 CAS-Nummer: 1799-97-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...