2-Chloro-6-methyl-4-(trifluoromethyl)pyridine 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC1938-1G
PC1938-5G
PC1938-10G
PC1938-250MG
APOSPC1938-1GEA
0
EUR
APOSPC1938-1G
APOSPC1938-5G
APOSPC1938-10G
APOSPC1938-250MG
2-Chloro-6-methyl-4-(trifluoromethyl)pyridine 97%
2-Chloro-6-methyl-4-(trifluoromethyl)pyridine
Formel:
C₇H₅ClF₃N Siedepunkt: 91…93 °C/69mmHg Lagertemperatur: Raumtemperatur |
MDL:
MFCD00052900 CAS-Nummer: 22123-14-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...