1,2,4-Tribrom-1,1,2-trifluorbutan 97%

Lieferant: Apollo Scientific

PC1956-5G
APOSPC1956-5GEA 170 EUR
APOSPC1956-5G
1,2,4-Tribrom-1,1,2-trifluorbutan 97%
1,2,4-Tribromo-1,1,2-trifluorobutane
Formel: C₄H₄Br₃F₃
Lagertemperatur: Raumtemperatur
CAS-Nummer: 2022-80-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR