Methyl-2,2-difluorobenzo[d][1,3]dioxole-5-carboxylate 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl 2,2-difluorobenzo[d][1,3]dioxole-5-carboxylate

PC19687-25G
APOSPC19687-25GEA 63.8 EUR
APOSPC19687-25G
Methyl-2,2-difluorobenzo[d][1,3]dioxole-5-carboxylate 98%
Methyl-2,2-difluorobenzo[d][1,3]dioxole-5-carboxylate
Formel: C₉H₆F₂O₄
Molecular Weight: 216,14 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06203771
CAS-Nummer: 773873-95-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR