1-Acetyl-3-methyl-4-nitro-5-(trifluoromethyl)-1H-pyrazole

Lieferant: Apollo Scientific
Achtung

PC1969-1G
APOSPC1969-1GEA 112 EUR
APOSPC1969-1G
1-Acetyl-3-methyl-4-nitro-5-(trifluoromethyl)-1H-pyrazole
1-Acetyl-3-methyl-4-nitro-5-(trifluoromethyl)-1H-pyrazole
Formel: C₇H₆F₃N₃O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD01320690
CAS-Nummer: 1017793-88-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR