3,5-Difluoro-2-hydrazinopyridine

Lieferant: Apollo Scientific
Achtung

PC1973-1G
APOSPC1973-1GEA 197 EUR
APOSPC1973-1G
3,5-Difluoro-2-hydrazinopyridine
3,5-Difluoro-2-hydrazinopyridine
Formel: C₅H₅F₂N₃
Lagertemperatur: Kühlschrank
MDL: MFCD09998072
CAS-Nummer: 851179-06-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR