Aluminium trifluoroacetate 97%

Lieferant: Apollo Scientific
Achtung

PC1976-1G PC1976-5G
APOSPC1976-1GEA 147 EUR
APOSPC1976-1G APOSPC1976-5G
Aluminium trifluoroacetate 97%
Aluminium trifluoroacetate
Formel: Al·3CF₃CO₂
Molecular Weight: 366.028 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03411227
CAS-Nummer: 36554-89-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR