1-Chloro-3,4-dibromo-3,4,4-trifluorobutane 97%

Lieferant: Apollo Scientific

PC1977-5G
APOSPC1977-5GEA 170 EUR
APOSPC1977-5G
1-Chloro-3,4-dibromo-3,4,4-trifluorobutane 97%
1-Chloro-3,4-dibromo-3,4,4-trifluorobutane
Formel: C₄H₄Br₂ClF₃
Molecular Weight: 304,331 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03094142
CAS-Nummer: 502457-70-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR