Silver(I) pentafluoropropanoate

Lieferant: Apollo Scientific
Gefahr

Synonyme: Pentafluoropropanoic acid silver(I) salt, Silver(I) pentafluoropropionate, Silver(I) perfluoropropanoate

PC1983-5G
APOSPC1983-5GEA 143 EUR
APOSPC1983-5G
Silver(I) pentafluoropropanoate
Silver(I) pentafluoropropanoate
Formel: C₃F₅O₂·Ag
Molecular Weight: 270,891 g/mol
Schmelzpunkt: 242…244 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00013251
CAS-Nummer: 509-09-1
EINECS: 208-093-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR