3-Chlor-5-(trifluormethyl)anilin 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Amino-5-chlor-α,α,α-trifluortoluol, 5-Chlor-α,α,α-trifluor-m-toluidin

PC1984-5G PC1984-1G
APOSPC1984-5GEA 24.8 EUR
APOSPC1984-5G APOSPC1984-1G
3-Chlor-5-(trifluormethyl)anilin 98%
3-Chlor-5-(trifluormethyl)anilin
Formel: C₇H₅ClF₃N
Molecular Weight: 195.57 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02684210
CAS-Nummer: 69411-05-8
EINECS: 273-989-1
UN: 2810
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR