1,3-Benzodioxol-5-yl-4,4,4-trifluorobutane-1,3-dione 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-[3,4-(Methylenedioxy)benzoyl]-1,1,1-trifluoroacetone

PC1987-1G
APOSPC1987-1GEA 102 EUR
APOSPC1987-1G
1,3-Benzodioxol-5-yl-4,4,4-trifluorobutane-1,3-dione 97%
1,3-Benzodioxol-5-yl-4,4,4-trifluorobutane-1,3-dione
Formel: C₁₁H₇F₃O₄
Molecular Weight: 260,166 g/mol
Schmelzpunkt: 104…108 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01570563
CAS-Nummer: 306935-39-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR