2-Amino-5-fluorbenzamid 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Fluoroanthranilamide

PC1994-1G PC1994-250MG PC1994-5G
APOSPC1994-1GEA 87.3 EUR
APOSPC1994-1G APOSPC1994-250MG APOSPC1994-5G
2-Amino-5-fluorbenzamid 97%
2-Amino-5-fluorbenzamid
Formel: C₇H₇FN₂O
Molecular Weight: 154.14 g/mol
Schmelzpunkt: 146…148 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03428522
CAS-Nummer: 63069-49-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR