2,4-Dimethylfluorobenzene

Lieferant: Apollo Scientific
Achtung

PC1997-25G PC1997-5G PC1997-100G
APOSPC1997-25GEA 0 EUR
APOSPC1997-25G APOSPC1997-5G APOSPC1997-100G
2,4-Dimethylfluorobenzene
2,4-Dimethylfluorobenzene
Formel: C₈H₉F
Lagertemperatur: Raumtemperatur
MDL: MFCD00060643
CAS-Nummer: 452-65-3
UN: 1993
ADR: 3,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR