2-Bromo-1-(diethoxymethyl)-4-fluorobenzene

Lieferant: Apollo Scientific
Achtung

PC200000-1G
APOSPC200000-1GEA 180 EUR
APOSPC200000-1G
2-Bromo-1-(diethoxymethyl)-4-fluorobenzene
2-Bromo-1-(diethoxymethyl)-4-fluorobenzene
Formel: C₁₁H₁₄BrFO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD22383957
CAS-Nummer: 198967-20-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR