1-Bromo-2-(chloromethyl)-4-fluorobenzene

Lieferant: Apollo Scientific
Achtung

PC200007-1G PC200007-500MG PC200007-5G
APOSPC200007-1GEA 168 EUR
APOSPC200007-1G APOSPC200007-500MG APOSPC200007-5G
1-Bromo-2-(chloromethyl)-4-fluorobenzene
1-Bromo-2-(chloromethyl)-4-fluorobenzene
Formel: C₇H₅BrClF
Lagertemperatur: Raumtemperatur
MDL: MFCD09263441
CAS-Nummer: 857276-61-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR