Ethyl-4-chloro-5-(trifluoromethyl)-pyridine-2-carboxylate
Lieferant: Apollo Scientific
|
Achtung
|
PC200013-1G
APOSPC200013-1GEA
1020
EUR
APOSPC200013-1G
Ethyl-4-chloro-5-(trifluoromethyl)-pyridine-2-carboxylate
Ethyl-4-chloro-5-(trifluoromethyl)-pyridine-2-carboxylate
Formel:
C₉H₇ClF₃NO₂ Molecular Weight: 253,61 g/mol Schmelzpunkt: 47…49 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD22573527 CAS-Nummer: 1407516-45-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...