3-Fluoro-6-methoxyquinoline

Lieferant: Apollo Scientific

PC200015-500MG
APOSPC200015-500MGEA 782 EUR
APOSPC200015-500MG
3-Fluoro-6-methoxyquinoline
3-Fluoro-6-methoxyquinoline
Formel: C₁₀H₈FNO
Lagertemperatur: Raumtemperatur
MDL: MFCD12400834
CAS-Nummer: 426842-85-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR