N-(4-Fluoro-2-methyl-6-nitrophenyl)acetamide

Lieferant: Apollo Scientific

PC200016-1G
APOSPC200016-1GEA 103 EUR
APOSPC200016-1G
N-(4-Fluoro-2-methyl-6-nitrophenyl)acetamide
N-(4-Fluoro-2-methyl-6-nitrophenyl)acetamide
Formel: C₉H₉FN₂O₃
Schmelzpunkt: 170…172 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD11520129
CAS-Nummer: 159020-73-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR