N-[3-(Trifluoromethyl)phenyl]-1,3,5-triazine-2,4-diamine

Lieferant: Apollo Scientific

PC200029-500MG
APOSPC200029-500MGEA 242 EUR
APOSPC200029-500MG
N-[3-(Trifluoromethyl)phenyl]-1,3,5-triazine-2,4-diamine
N-[3-(Trifluoromethyl)phenyl]-1,3,5-triazine-2,4-diamine
Formel: C₁₀H₈F₃N₅
Molecular Weight: 255,2 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00179648
CAS-Nummer: 3832-69-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR