Ethyl-3-fluoropyridine-2-carboxylate 95%
Lieferant: Apollo Scientific
|
Achtung
|
PC200030-1G
PC200030-500MG
PC200030-5G
APOSPC200030-1GEA
0
EUR
APOSPC200030-1G
APOSPC200030-500MG
APOSPC200030-5G
Ethyl-3-fluoropyridine-2-carboxylate 95%
Ethyl-3-fluoropyridine-2-carboxylate
Formel:
C₈H₈FNO₂ Schmelzpunkt: 74…76 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD11616884 CAS-Nummer: 1187732-69-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...