Methyl 2-amino-2-[4-(trifluoromethyl)phenyl]acetate

Lieferant: Apollo Scientific

PC200032-500MG
APOSPC200032-500MGEA 170 EUR
APOSPC200032-500MG
Methyl 2-amino-2-[4-(trifluoromethyl)phenyl]acetate
Methyl 2-amino-2-[4-(trifluoromethyl)phenyl]acetate
Formel: C₁₀H₁₀F₃NO₂
Molecular Weight: 233,187 g/mol
Schmelzpunkt: 50…52 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD12151721
CAS-Nummer: 439088-62-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR