2,3,5-Trichloro-4,6-bistrifluoromethyl pyridine

Lieferant: Apollo Scientific

PC200035-500MG
APOSPC200035-500MGEA 545 EUR
APOSPC200035-500MG
2,3,5-Trichloro-4,6-bistrifluoromethyl pyridine
2,3,5-Trichloro-4,6-bistrifluoromethyl pyridine
Formel: C₇Cl₃F₆N
Molecular Weight: 318,431 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22689161
CAS-Nummer: 1420537-83-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR