Methyl-3-cyan-5-fluorbenzoat
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Methyl 3-cyano-5-fluorobenzoate
PC200038-1G
APOSPC200038-1GEA
40.6
EUR
APOSPC200038-1G
Methyl-3-cyan-5-fluorbenzoat
Methyl-3-cyan-5-fluorbenzoat
Formel:
C₉H₆FNO₂ Molecular Weight: 179,15 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08688582 CAS-Nummer: 886732-29-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...