7-Chloro-6-fluoro-1-methoxyisoquinoline
Lieferant: Apollo Scientific
PC200044-1G
PC200044-500MG
APOSPC200044-1GEA
538
EUR
APOSPC200044-1G
APOSPC200044-500MG
7-Chloro-6-fluoro-1-methoxyisoquinoline
7-Chloro-6-fluoro-1-methoxyisoquinoline
Formel:
C₁₀H₇ClFNO Molecular Weight: 211.62 g/mol |
MDL:
MFCD18374925 CAS-Nummer: 1202006-81-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...